Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ADAM19 Rabbit anti-Human, Polyclonal Antibody, Abnova™

Catalog No. 89121358 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-121-358 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-121-358 Supplier Abnova Corporation Supplier No. PAB19828
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of ADAM19.

This gene encodes a member of the ADAM (a disintegrin and metalloprotease domain) family. Members of this family are membrane-anchored proteins structurally related to snake venom disintegrins, and have been implicated in a variety of biological processes involving cell-cell and cell-matrix interactions, including fertilization, muscle development, and neurogenesis. This member is a type I transmembrane protein and serves as a marker for dendritic cell differentiation. It has also been demonstrated to be an active metalloproteinase, which may be involved in normal physiological and pathological processes such as cells migration, cell adhesion, cell-cell and cell-matrix interactions, and signal transduction. [provided by RefSeq

Sequence: REPGWTSKGSEEGSC

Specifications

Antigen ADAM19
Applications ELISA, Immunohistochemistry (PFA fixed), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of ADAM19.
Dilution ELISA (1:10000-1:40000) Western Blot (1:200-500) The optimal working dilution should be determined by the end user.
Formulation In serum (0.02% sodium azide)
Gene ADAM19
Gene Accession No. Q9H013
Gene Alias FKSG34/MADDAM/MLTNB
Gene Symbols ADAM19
Host Species Rabbit
Immunogen A synthetic peptide corresponding to 15 amino acids near N-terminus of human ADAM19.
Purification Method Protein A purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8728
Target Species Human
Content And Storage Store at 4°C for three months. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.