Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ADAM30, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89120387 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-120-387 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-120-387 Supplier Abnova Corporation Supplier No. PAB18828
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of ADAM30.

This gene encodes a member of the ADAM (a disintegrin and metalloprotease domain) family. Members of this family are membrane-anchored proteins structurally related to snake venom disintegrins, and have been implicated in a variety of biological processes involving cell-cell and cell-matrix interactions, including fertilization, muscle development, and neurogenesis. This gene is testis-specific and contains a polymorphic region, resulting in isoforms with varying numbers of C-terminal repeats. [provided by RefSeq

Specifications

Antigen ADAM30
Applications ELISA, Immunohistochemistry (PFA fixed)
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of ADAM30.
Dilution ELISA (1:1000-1:10000) Immunohistochemistry (1:50-1:200) The optimal working dilution should be determined by the end user.
Formulation In buffer containing 0.02% sodium azide, 2% BSA
Gene ADAM30
Gene Alias svph4
Gene Symbols ADAM30
Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids at internal region of human ADAM30.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11085
Target Species Human
Content And Storage Store at 4°C for three months. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.