Learn More
ADAMTS12, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant ADAMTS12.
Supplier: Abnova Corporation PAB22935
Description
This gene encodes a member of the ADAMTS (a disintegrin and metalloproteinase with thrombospondin motifs) protein family. Members of the family share several distinct protein modules, including a propeptide region, a metalloproteinase domain, a disintegrin-like domain, and a thrombospondin type 1 (TS-1) motif. Individual members of this family differ in the number of C-terminal TS-1 motifs, and some have unique C-terminal domains. The enzyme encoded by this gene contains eight TS-1 motifs. It may play roles in pulmonary cells during fetal development or in tumor processes through its proteolytic activity or as a molecule potentially involved in regulation of cell adhesion. [provided by RefSeq
Sequence: SSDTGPTSEGGLVATTTSGSGLSSSRNPITWPVTPFYNTLTKGPEMEIHSGSGEEREQPEDKDESNPVIWTKIRVPGNDAPVESTEMPLAPPLTPDLSRESWWPPFSTVMEGLLPSQRPTTSETGTPRVEGMVTEKPANTLLPLSpecifications
ADAMTS12 | |
Polyclonal | |
Rabbit polyclonal antibody raised against recombinant ADAMTS12. | |
In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
P58397 | |
ADAMTS12 | |
Recombinant protein corresponding to amino acids of human ADAMTS12. | |
100 μL | |
Primary | |
Human | |
Liquid |
Immunohistochemistry (PFA fixed) | |
Unconjugated | |
Immunohistochemistry (1:50-1:200) The optimal working dilution should be determined by the end user. | |
ADAMTS12 | |
PRO4389 | |
Rabbit | |
Antigen affinity purification | |
RUO | |
81792 | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only