Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ADAMTS2, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89120375 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-120-375 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-120-375 Supplier Abnova Corporation Supplier No. PAB18817
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of ADAMTS2.

This gene encodes a member of the ADAMTS (a disintegrin and metalloproteinase with thrombospondin motifs) protein family. Members of the family share several distinct protein modules, including a propeptide region, a metalloproteinase domain, a disintegrin-like domain, and a thrombospondin type 1 (TS) motif. Individual members of this family differ in the number of C-terminal TS motifs, and some have unique C-terminal domains. The enzyme encoded by this gene excises the N-propeptide of type I, type II and type V procollagens. Mutations in this gene cause Ehlers-Danlos syndrome type VIIC, a recessively inherited connective-tissue disorder. Alternative splicing results in two transcript variants. The short transcript encodes a protein which has no significant procollagen N-peptidase activity. [provided by RefSeq

Specifications

Antigen ADAMTS2
Applications ELISA, Immunohistochemistry (PFA fixed), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of ADAMTS2.
Dilution ELISA (1:1000-1:10000) Western Blot (1:200-1:1000) Immunohistochemistry (1:100-1:500) The optimal working dilution should be determined by the end user.
Formulation In buffer containing 0.02% sodium azide, 0.5% BSA
Gene ADAMTS2
Gene Alias ADAM-TS2/ADAMTS-3/NPI/PCINP/PCPNI/hPCPNI
Gene Symbols ADAMTS2
Host Species Rabbit
Immunogen A synthetic peptide corresponding to 16 amino acids at C-terminus of human ADAMTS2.
Purification Method Peptide affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9509
Target Species Human
Content And Storage Store at 4°C for three months. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.