Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ADAMTSL1, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89120380 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-120-380 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-120-380 Supplier Abnova Corporation Supplier No. PAB18821
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of ADAMTSL1.

This gene encodes a secreted protein and member of the ADAMTS (a disintegrin and metalloproteinase with thrombospondin motif) family. This protein lacks the metalloproteinase and disintegrin-like domains, which are typical of the ADAMTS family, but contains other ADAMTS domains, including the thrombospondin type 1 motif. This protein may have important functions in the extracellular matrix. Alternative splicing results in multiple transcript variants encoding distinct proteins. [provided by RefSeq

Specifications

Antigen ADAMTSL1
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of ADAMTSL1.
Dilution ELISA (1:160000) Western Blot (1:1000-1:2000) The optimal working dilution should be determined by the end user.
Formulation In buffer containing 0.02% sodium azide
Gene ADAMTSL1
Gene Alias ADAMTSR1/C9orf94/DKFZp686L03130/FLJ35283/FLJ41032/FLJ46891/MGC118803/MGC118805/MGC40193/PUNCTIN
Gene Symbols ADAMTSL1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids at C-terminus of human ADAMTSL1.
Purification Method Protein A purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 92949
Target Species Human
Content And Storage Store at 4°C for three months. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.