Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ADCY2 Rabbit anti-Human, Polyclonal Antibody, Abnova™

Catalog No. 89121346 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-121-346 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-121-346 Supplier Abnova Corporation Supplier No. PAB19816
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of ADCY2.

This gene encodes a member of the family of adenylate cyclases, which are membrane-associated enzymes that catalyze the formation of the secondary messenger cyclic adenosine monophosphate (cAMP). This enzyme is insensitive to Ca(2+)/calmodulin, and is stimulated by the G protein beta and gamma subunit complex. [provided by RefSeq]

Specifications

Antigen ADCY2
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of ADCY2.
Dilution ELISA (1:160000) Western Blot (1:200-500) The optimal working dilution should be determined by the end user.
Formulation In serum (0.02% sodium azide)
Gene ADCY2
Gene Accession No. Q08462
Gene Alias AC2/FLJ16822/FLJ45092/HBAC2/KIAA1060/MGC133314
Gene Symbols ADCY2
Host Species Rabbit
Immunogen A synthetic peptide corresponding to 15 amino acids at N-terminus of human ADCY2.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 108
Target Species Human
Content And Storage Store at 4°C for three months. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.