Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ADCY9, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89121271 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-121-271 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-121-271 Supplier Abnova Corporation Supplier No. PAB19743
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of ADCY9.

Adenylate cyclase is a membrane bound enzyme that catalyses the formation of cyclic AMP from ATP. It is regulated by a family of G protein-coupled receptors, protein kinases, and calcium. The type 9 adenylyl cyclase is a widely distributed adenylyl cyclase, and it is stimulated by beta-adrenergic receptor activation but is insensitive to forskolin, calcium, and somatostatin. [provided by RefSeq

Specifications

Antigen ADCY9
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of ADCY9.
Dilution ELISA (1:160000) Western Blot (1:200-500) The optimal working dilution should be determined by the end user.
Formulation In serum (0.02% sodium azide)
Gene ADCY9
Gene Accession No. O60503
Gene Alias AC9
Gene Symbols ADCY9
Host Species Rabbit
Immunogen A synthetic peptide corresponding to 14 amino acids at N-terminus of human ADCY9.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 115
Target Species Human
Content And Storage Store at 4°C for three months. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.