Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ADD1/ADD2, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of ADD1/ADD2.
Supplier: Abnova Corporation PAB27016
Specifications
ADD1/ADD2 | |
Polyclonal | |
Unconjugated | |
Western Blot (1:500-1:1000) Immunohistochemistry (1:50-1:200) Immunofluorescence (1:50-1:200) Immunoprecipitor (1:50-1:200) The optimal working dilution should be determined by the end user. | |
P35611 (Gene ID : 118);P35612 (Gene ID : 119) | |
ADD1 | |
A synthetic peptide corresponding to human ADD1/ADD2. | |
100 μL | |
Primary | |
ADD1/ADD2 polyclonal antibody detects endogenous levels of ADD1/ADD2 protein. | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
ELISA, Immunofluorescence, Immunohistochemistry (PFA fixed), Immunoprecipitation, Western Blot | |
1 mg/mL | |
Rabbit polyclonal antibody raised against synthetic peptide of ADD1/ADD2. | |
In PBS, pH 7.2 (0.05% sodium azide) | |
ADDA/MGC3339/MGC44427 | |
Rabbit | |
Antigen affinity purification | |
RUO | |
118/119 | |
Human, Mouse, Rat | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction