Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ADIPOQ Rabbit anti-Human, Polyclonal Antibody, Abnova™

Catalog No. 89116826 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-116-826 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-116-826 Supplier Abnova Corporation Supplier No. PAB13761

Rabbit polyclonal antibody raised against recombinant ADIPOQ.

This gene is expressed in adipose tissue exclusively. It encodes a protein with similarity to collagens X and VIII and complement factor C1q. The encoded protein circulates in the plasma and is involved with metabolic and hormonal processes. [provided by RefSeq

Specifications

Antigen ADIPOQ
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against recombinant ADIPOQ.
Dilution ELISA (1:5000-1:10000) Western Blot (1:5000-1:10000) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4
Gene ADIPOQ
Gene Alias ACDC/ACRP30/ADPN/APM-1/APM1/GBP28/adiponectin
Gene Symbols ADIPOQ
Host Species Rabbit
Immunogen Recombinant protein corresponding to human ADIPOQ.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9370
Test Specificity Recognizes human adiponectin. Detects a band of ∼30 KDa by Western blot.
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.