Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ADIPOQ Rabbit anti-Human, Polyclonal Antibody, Abnova™

Catalog No. 89131694 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
150 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-131-694 150 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-131-694 Supplier Abnova Corporation Supplier No. PAB5030
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of ADIPOQ.

This gene is expressed in adipose tissue exclusively. It encodes a protein with similarity to collagens X and VIII and complement factor C1q. The encoded protein circulates in the plasma and is involved with metabolic and hormonal processes. [provided by RefSeq

Sequence: CLPGHDGETTTQGPGVL

Specifications

Antigen ADIPOQ
Applications ELISA, Immunoprecipitation
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of ADIPOQ.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.5 (50% glycerol, 0.01% thimerosal)
Gene ADIPOQ
Gene Alias ACDC/ACRP30/ADPN/APM-1/APM1/GBP28/adiponectin
Gene Symbols ADIPOQ
Host Species Rabbit
Immunogen A synthetic peptide (conjugated with KLH) corresponding to human ADIPOQ.
Quantity 150 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9370
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.