Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ADIPOR1 Rabbit anti-Human, Polyclonal Antibody, Abnova™

Catalog No. 89116809 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-116-809 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-116-809 Supplier Abnova Corporation Supplier No. PAB13695
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of ADIPOR1.

The adiponectin receptors, ADIPOR1 and ADIPOR2 (MIM 607946), serve as receptors for globular and full-length adiponectin (MIM 605441) and mediate increased AMPK (see MIM 602739) and PPAR-alpha (PPARA; MIM 170998) ligand activities, as well as fatty acid oxidation and glucose uptake by adiponectin (Yamauchi et al., 2003 [PubMed 12802337]).[supplied by OMIM

Sequence: SSHKGSVVAQGNGAPASNREADTVELAELGPLLEE

Specifications

Antigen ADIPOR1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of ADIPOR1.
Dilution Western Blot (1:1000) The optimal working dilution should be determined by the end user.
Formulation In serum (0.02% sodium azide)
Gene ADIPOR1
Gene Alias ACDCR1/CGI-45/CGI45/FLJ25385/FLJ42464/PAQR1/TESBP1A
Gene Symbols ADIPOR1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 2-36 of human ADIPOR1.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51094
Test Specificity Recognizes human adiponectin receptor 1.
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.