Learn More
ADORA3 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of ADORA3.
Supplier: Abnova Corporation PAB24973
Description
This gene encodes a protein that belongs to the family of adenosine receptors, which are G-protein-coupled receptors that are involved in a variety of intracellular signaling pathways and physiological functions. The receptor encoded by this gene mediates a sustained cardioprotective function during cardiac ischemia, it is involved in the inhibition of neutrophil degranulation in neutrophil-mediated tissue injury, it has been implicated in both neuroprotective and neurodegenerative effects, and it may also mediate both cell proliferation and cell death. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq
Specifications
ADORA3 | |
Polyclonal | |
Unconjugated | |
Western Blot (1:500-1:1000) Immunohistochemistry (1:50-1:200) Immunofluorescence (1:50-1:200) The optimal working dilution should be determined by the end user. | |
ADORA3 | |
ADORA3 | |
A synthetic peptide corresponding to ADORA3. | |
100 μL | |
Primary | |
ADORA3 polyclonal antibody detects endogenous levels of Adenosine ADORA3 protein. | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Immunofluorescence, Immunohistochemistry, Western Blot | |
1 mg/mL | |
Rabbit polyclonal antibody raised against synthetic peptide of ADORA3. | |
In PBS, pH 7.2 (0.05% sodium azide) | |
A3AR/AD026/bA552M11.5 | |
Rabbit | |
Antigen affinity purification | |
RUO | |
140 | |
Human | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only