Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ADRA1B Rabbit anti-Human, Polyclonal Antibody, Abnova™

Catalog No. 89121334 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-121-334 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-121-334 Supplier Abnova Corporation Supplier No. PAB19804
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of ADRA1B.

Alpha-1-adrenergic receptors (alpha-1-ARs) are members of the G protein-coupled receptor superfamily. They activate mitogenic responses and regulate growth and proliferation of many cells. There are 3 alpha-1-AR subtypes: alpha-1A, -1B and -1D, all of which signal through the Gq/11 family of G-proteins and different subtypes show different patterns of activation. This gene encodes alpha-1B-adrenergic receptor, which induces neoplastic transformation when transfected into NIH 3T3 fibroblasts and other cell lines. Thus, this normal cellular gene is identified as a protooncogene. This gene comprises 2 exons and a single large intron of at least 20kb that interrupts the coding region. [provided by RefSeq

Specifications

Antigen ADRA1B
Applications ELISA, Immunohistochemistry (PFA fixed), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of ADRA1B.
Dilution ELISA (1:1000-1:10000) Western Blot (1:200-1000) Immunohistochemistry (1:100-500.) The optimal working dilution should be determined by the end user.
Formulation In serum (1% BSA, 0.02% sodium azide)
Gene ADRA1B
Gene Accession No. P35368
Gene Alias ADRA1/ALPHA1BAR
Gene Symbols ADRA1B
Host Species Rabbit
Immunogen A synthetic peptide corresponding to 15 amino acids at C-terminus of human ADRA1B.
Purification Method Peptide affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 147
Target Species Human
Content And Storage Store at 4°C for three months. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.