Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ADRA1D Rabbit anti-Fruit fly, Human, Mouse, Rat, Zebra fish, Polyclonal Antibody, Abnova™

Catalog No. 89121276 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-121-276 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-121-276 Supplier Abnova Corporation Supplier No. PAB19748
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of ADRA1D.

Alpha-1-adrenergic receptors (alpha-1-ARs) are members of the G protein-coupled receptor superfamily. They activate mitogenic responses and regulate growth and proliferation of many cells. There are 3 alpha-1-AR subtypes: alpha-1A, -1B and -1D, all of which signal through the Gq/11 family of G-proteins and different subtypes show different patterns of activation. This gene encodes alpha-1D-adrenergic receptor. Similar to alpha-1B-adrenergic receptor gene, this gene comprises 2 exons and a single intron that interrupts the coding region. [provided by RefSeq

Specifications

Antigen ADRA1D
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of ADRA1D.
Dilution ELISA (1:10000-1:40000) Western Blot (1:500-2000) The optimal working dilution should be determined by the end user.
Formulation In serum (0.02% sodium azide)
Gene ADRA1D
Gene Accession No. P25100
Gene Alias ADRA1/ADRA1A/ADRA1R/ALPHA1/DAR/dJ779E11.2
Gene Symbols ADRA1D
Host Species Rabbit
Immunogen A synthetic peptide corresponding to internal region of human ADRA1D.
Purification Method Protein A purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 146
Target Species Drosophila, Fish, Human, Mouse, Rat
Content And Storage Store at 4°C for three months. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.