Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AGGF1 Rabbit anti-Human, Polyclonal Antibody, Abnova™

Catalog No. 89115869 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-115-869 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-115-869 Supplier Abnova Corporation Supplier No. PAB12494
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of AGGF1.

The AGGF1 gene encodes a potent angiogenic factor that contains a forkhead-associated domain and a G-patch domain (Tian et al., 2004 [PubMed 14961121]).[supplied by OMIM

Specifications

Antigen AGGF1
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of AGGF1.
Dilution Immunohistochemistry (1:250) Western Blot (1:500) The optimal working dilution should be determined by the end user.
Formulation In antiserum
Gene AGGF1
Gene Accession No. Q8N302
Gene Alias FLJ10283/GPATC7/GPATCH7/HSU84971/HUS84971/VG5Q
Gene Symbols AGGF1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 500-600 of human AGGF1.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55109
Test Specificity This antibody is specific to human VG5Q.
Target Species Human
Content And Storage Store at 4°C for short term. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.