Learn More
AGL, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of AGL.
Supplier: Abnova Corporation PAB4298
Description
This gene encodes the glycogen debrancher enzyme which is involved in glycogen degradation. This enzyme has two independent catalytic activities which occur at different sites on the protein: a 4-alpha-glucotransferase activity and a amylo-1,6-glucosidase activity. Mutations in this gene are associated with glycogen storage disease although a wide range of enzymatic and clinical variability occurs which may be due to tissue-specific alternative splicing. Alternatively spliced transcripts encoding different isoforms have been described. [provided by RefSeq
Specifications
AGL | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of AGL. | |
In PBS (0.09% sodium azide) | |
NP_000019;P35573 | |
AGL | |
A synthetic peptide (conjugated with KLH) corresponding to C-terminus of human AGL. | |
400 μL | |
Primary | |
Human | |
Liquid |
ELISA, Immunofluorescence, Western Blot | |
Unconjugated | |
ELISA (1:1000) Western Blot (1:100-500) The optimal working dilution should be determined by the end user. | |
AGL | |
GDE | |
Rabbit | |
Protein A purification | |
RUO | |
178 | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only