Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AGPAT9, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89123991 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-123-991 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-123-991 Supplier Abnova Corporation Supplier No. PAB22285
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against recombinant AGPAT9.

Glycerol-3-phosphate (G3P) acyltransferases (GPAT; EC 2.3.1.15), such as GPAM (MIM 602395) and GPAT3, catalyze the initial step of de novo triacylglycerol (TAG) synthesis by converting glycerol-3-phosphate (G3P) to lysophosphatidic acid (LPA) (Cao et al., 2006 [PubMed 17170135]).[supplied by OMIM

Sequence: ILVKTLEWATIRIEKGTPKESILKNSASVGIIQRDESPMEKGLSGLRGRDFELSDVFYFSKKGLEAIVEDEVTQRFSSEELVSWNLLTRTN

Specifications

Antigen AGPAT9
Applications Immunohistochemistry (PFA fixed), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against recombinant AGPAT9.
Dilution Immunohistochemistry (1:500-1:1000) Western Blot (1:250-1:500) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide)
Gene AGPAT9
Gene Accession No. Q53EU6
Gene Alias AGPAT8/GPAT3/HMFN0839/LPAAT-theta/MAG1/MGC11324
Gene Symbols AGPAT9
Host Species Rabbit
Immunogen Recombinant protein corresponding to amino acids of human AGPAT9.
Purification Method Antigen affinity purification
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84803
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.