Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AGR2 Rabbit anti-Bovine, Human, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89115866 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-115-866 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-115-866 Supplier Abnova Corporation Supplier No. PAB12491
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of AGR2.

Specifications

Antigen AGR2
Applications Immunohistochemistry (PFA fixed)
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of AGR2.
Dilution Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:100) The optimal working dilution should be determined by the end user.
Formulation In PBS (30% glycerol, 0.09% sodium azide)
Gene AGR2
Gene Accession No. O95994
Gene Alias AG2/GOB-4/HAG-2/XAG-2
Gene Symbols AGR2
Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 50-100 of human AGR2.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10551
Target Species Bovine, Human, Mouse
Content And Storage Store at 4°C for short term. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.