Learn More
AKAP10, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of AKAP10.
Supplier: Abnova Corporation PAB19746
Description
The A-kinase anchor proteins (AKAPs) are a group of structurally diverse proteins, which have the common function of binding to the regulatory subunit of protein kinase A (PKA) and confining the holoenzyme to discrete locations within the cell. This gene encodes a member of the AKAP family. The encoded protein interacts with both the type I and type II regulatory subunits of PKA; therefore, it is a dual-specific AKAP. This protein is highly enriched in mitochondria. It contains RGS (regulator of G protein signalling) domains, in addition to a PKA-RII subunit-binding domain. The mitochondrial localization and the presence of RGS domains may have important implications for the function of this protein in PKA and G protein signal transduction. [provided by RefSeq
Specifications
AKAP10 | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of AKAP10. | |
In serum (0.02% sodium azide) | |
O43572 | |
AKAP10 | |
A synthetic peptide corresponding to 14 amino acids at C-terminus of human AKAP10. | |
RUO | |
11216 | |
Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
ELISA, Western Blot | |
Unconjugated | |
ELISA (1:160000) Western Blot (1:500-1000) The optimal working dilution should be determined by the end user. | |
AKAP10 | |
D-AKAP2/MGC9414/PRKA10 | |
Rabbit | |
100 μg | |
Primary | |
Human | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only