Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AKR7A2, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against partial recombinant AKR7A2.
Supplier: Abnova Corporation PAB18833
Description
Aldo-keto reductases, such as AKR7A2, are involved in the detoxification of aldehydes and ketones.[supplied by OMIM
Specifications
AKR7A2 | |
Polyclonal | |
Rabbit polyclonal antibody raised against partial recombinant AKR7A2. | |
In buffer containing 0.02% sodium azide | |
AFAR/AFAR1/AFB1-AR1/AKR7 | |
Rabbit | |
100 μg | |
Primary | |
Human, Rat | |
Liquid |
ELISA, Immunohistochemistry (PFA fixed), Western Blot | |
Unconjugated | |
ELISA (1:10000-1:80000) Western Blot (1:200-1:1000) Immunohistochemistry (1:50-1:200) The optimal working dilution should be determined by the end user. | |
AKR7A2 | |
AKR7A2 | |
Recombinant protein corresponding to amino acids 184-329 of human AKR7A2. | |
RUO | |
8574 | |
Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction