Learn More
AKT1 Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Shop All Abnova Corporation ProductsDescription
The serine-threonine protein kinase encoded by the AKT1 gene is catalytically inactive in serum-starved primary and immortalized fibroblasts. AKT1 and the related AKT2 are activated by platelet-derived growth factor. The activation is rapid and specific, and it is abrogated by mutations in the pleckstrin homology domain of AKT1. It was shown that the activation occurs through phosphatidylinositol 3-kinase. In the developing nervous system AKT is a critical mediator of growth factor-induced neuronal survival. Survival factors can suppress apoptosis in a transcription-independent manner by activating the serine/threonine kinase AKT1, which then phosphorylates and inactivates components of the apoptotic machinery. Multiple alternatively spliced transcript variants have been found for this gene. [provided by RefSeq
Specifications
Specifications
| Antigen | AKT1 |
| Applications | ELISA, Immunohistochemistry, Immunohistochemistry (PFA fixed), Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Description | Rabbit polyclonal antibody raised against synthetic peptide of AKT1. |
| Dilution | Western Blot (1:500∼1:1000) Immunohistochemistry (1:50∼1:100) ELISA(1:4000) The optimal working dilution should be determined by the end user. |
| Formulation | In PBS, 150mM NaCl, pH 7.4 (50% glycerol, 0.02% sodium azide) |
| Gene | AKT1 |
| Gene Accession No. | P31749 |
| Gene Alias | AKT/MGC99656/PKB/PKB-ALPHA/PRKBA/RAC/RAC-ALPHA |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.