Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ALB Rabbit anti-Bovine, Polyclonal Antibody, Abnova™

Catalog No. 89117589 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-117-589 2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-117-589 Supplier Abnova Corporation Supplier No. PAB15362
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against native ALB.

Specifications

Antigen ALB
Applications ELISA, Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against native ALB.
Dilution The optimal working dilution should be determined by the end user.
Formulation In buffer containing 0.05% sodium azide
Gene ALB
Gene Alias MGC179277
Gene Symbols ALB
Host Species Rabbit
Immunogen Native purified bovine ALB.
Quantity 2 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 280717
Test Specificity This antibody shows a single band of precipitation with whole bovine serum and purified BSA. The antiserum has not been assayed for interspecies cross-reactivity.
Target Species Bovine
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.