Learn More
ALDH1A1 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of ALDH1A1.
Supplier: Abnova Corporation PAB3123
Description
This protein belongs to the aldehyde dehydrogenases family of proteins. Aldehyde dehydrogenase is the second enzyme of the major oxidative pathway of alcohol metabolism. Two major liver isoforms of this enzyme, cytosolic and mitochondrial, can be distinguished by their electrophoretic mobilities, kinetic properties, and subcellular localizations. Most Caucasians have two major isozymes, while approximately 50% of Orientals have only the cytosolic isozyme, missing the mitochondrial isozyme. A remarkably higher frequency of acute alcohol intoxication among Orientals than among Caucasians could be related to the absence of the mitochondrial isozyme. This gene encodes a cytosolic isoform, which has a high affinity for aldehydes. [provided by RefSeq
Specifications
ALDH1A1 | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of ALDH1A1. | |
In PBS (0.09% sodium azide) | |
NP_000680;P00352 | |
ALDH1A1 | |
A synthetic peptide (conjugated with KLH) corresponding to internal region of human ALDH1A1. | |
400 μL | |
Primary | |
Human | |
Liquid |
ELISA, Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Western Blot (1:1000) Immunofluorescence (1:10-50) Immunohistochemistry (1:50) Flow cytometry (1:10-50) The optimal working dilution should be determined by the end user. | |
ALDH1A1 | |
ALDC/ALDH-E1/ALDH1/ALDH11/MGC2318/PUMB1/RALDH1 | |
Rabbit | |
Protein A purification | |
RUO | |
216 | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only