Learn More
ALDH1A3, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant ALDH1A3.
Supplier: Abnova Corporation PAB24490
Description
Aldehyde dehydrogenase isozymes are thought to play a major role in the detoxification of aldehydes generated by alcohol metabolism and lipid peroxidation. The enzyme encoded by this gene uses retinal as a substrate, either in a free or cellular retinol-binding protein form. [provided by RefSeq
Sequence: ERGGAMATANGAVENGQPDRKPPALPRPIRNLEVKFTKIFINSpecifications
| ALDH1A3 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant ALDH1A3. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| P47895 | |
| ALDH1A3 | |
| Recombinant protein corresponding to amino acids of human ALDH1A3. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
| Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Immunohistochemistry (1:20-1:50) Western Blot (1:250-1:500) The optimal working dilution should be determined by the end user. | |
| ALDH1A3 | |
| ALDH1A6/ALDH6/RALDH3 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 220 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only