Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ALG13, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89130112 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-130-112 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-130-112 Supplier Abnova Corporation Supplier No. PAB28600
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against recombinant ALG13.

Asparagine (N)-glycosylation is an essential modification that regulates protein folding and stability. ALG13 and ALG14 (MIM 612866) constitute the UDP-GlcNAc transferase, which catalyzes a key step in endoplasmic reticulum N-linked glycosylation (Averbeck et al., 2007 [PubMed 17686769]).[supplied by OMIM

Sequence: VGTTSFDDLIACVSAPDSLQKIESLGYNRLILQIGRGTVVPEPFSTESFTLDVYRYKDSLKEDIQKADLVISHAGAGSCLETLEKGKPLVVVINEKLMNNHQLELAKQLHKEGHLFYCTCSTLPGLLQSMDLSTLKCYPPG

Specifications

Antigen ALG13
Applications Immunohistochemistry (PFA fixed), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against recombinant ALG13.
Dilution Western Blot (1:100-1:250) Immunohistochemistry (1:200-1:500) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.2 (40% glycerol, 0.02% sodium azide)
Gene ALG13
Gene Accession No. Q9NP73
Gene Alias CXorf45/FLJ23018/GLT28D1/MDS031/MGC12423/YGL047W
Gene Symbols ALG13
Host Species Rabbit
Immunogen Recombinant protein corresponding to amino acids of human ALG13.
Purification Method Antigen affinity purification
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79868
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.