Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Rabbit anti-ALKBH1, Polyclonal,
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma™ ABE1310
Description
Specifically detects ALKBH1 in Human, Mouse samples, and it is validated for Immunocytochemistry, Immunohistochemistry
ALKBH1 is a DNA lyase that is critical for DNA repair and specifically repairs alkylated single-stranded DNA and RNA containing 3-methylcytosine by oxidative demethylation. ALKBH1 can also cleave both single-stranded and double-stranded DNA at basic sites, and ALKBH1 can induce double stranded breaks at basic sites. Finally, ALKBH1 plays a role in placental trophoblast lineage differentiation during development, and ALKBH1 may also play a role in neuronal development and migration. ALKBH1 is widely expressed and localizes to mitochondria and nucleus. The nuclear localization of ALKBH1 appears to be as part of fusion proteins with nuclear targeted proteins or when the first 26 N-terminal residues are removed.
Specifications
ALKBH1 | |
Polyclonal | |
Q13686 | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Human, Mouse |
Immunocytochemistry, Immunohistochemistry | |
Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide. | |
ALKBH1; ABH; ABH1; ALKBH | |
KLH-conjugated linear peptide corresponding to the substrate binding region of human ALKBH1. | |
100 μg | |
Epigenetics and Nuclear Function | |
NP_006011 | |
Stable for one year at 2°C to 8°C from date of receipt. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction