Learn More
ALOX15B, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant ALOX15B .
Supplier: Abnova Corporation PAB13608
Description
This gene encodes a member of the lipoxygenase family of structurally related nonheme iron dioxygenases involved in the production of fatty acid hydroperoxides. The encoded protein converts arachidonic acid exclusively to 15S-hydroperoxyeicosatetraenoic acid, while metabolizing linoleic acid less effectively. This gene is located in a cluster of related genes and a pseudogene that spans approximately 100 kilobases on the short arm of chromosome 17. Alternatively spliced transcript variants encoding different isoforms have been described. [provided by RefSeq
Specifications
ALOX15B | |
Polyclonal | |
Rabbit polyclonal antibody raised against recombinant ALOX15B . | |
ALOX15B | |
ALOX15B | |
Recombinant protein corresponding to human ALOX15B . | |
RUO | |
247 | |
Human, Mouse | |
Liquid |
ELISA, Immunohistochemistry (PFA fixed), Immunoprecipitation, Western Blot | |
Unconjugated | |
Western Blot (1:1000) The optimal working dilution should be determined by the end user. | |
15-LOX-2 | |
Rabbit | |
100 μL | |
Primary | |
This rabbit antibody reacts specifically with 15-lipoxygenase form-2 in human tissues and 8-lipoxygenase in murine tissues. 15-Lipoxygenase Form-2 (15-LOX-2) is expressed primarily in lung tissue, hair roots, the prostate and the cornea and is a useful biomarker in eicosanoid studies. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only