Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alox8, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89116763 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-116-763 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-116-763 Supplier Abnova Corporation Supplier No. PAB13607
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against Alox8.

Specifications

Antigen Alox8
Applications ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against Alox8.
Dilution Western Blot (1:3000) The optimal working dilution should be determined by the end user.
Formulation In unpurified antiserum.
Gene Alox8
Gene Alias 8-LOX/8S-LOX/Alox15b
Gene Symbols Alox8
Host Species Rabbit
Immunogen Mouse tissues Alox8.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11688
Test Specificity This rabbit antibody is specific to 8-lipoxygenase in mice. In human tissues, it is specific to 15-lipoxygenase form-2.
Target Species Human, Mouse
Content And Storage Store at -20°C. For long term storage store at -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.