Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AMACR Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89114925 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-114-925 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-114-925 Supplier Abnova Corporation Supplier No. PAB1121

Rabbit polyclonal antibody raised against synthetic peptide of AMACR.

Alpha-methylacyl-CoA racemase is responsible for the conversion of pristanoyl-CoA and C27-bile acyl-CoAs to their (S)-stereoisomers, which are the only stereoisomers that can be degraded via peroxisomal beta-oxidation.[supplied by OMIM

Sequence: RVDRPGSRYDVSRLGR

Specifications

Antigen AMACR
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description alpha-methylacyl-CoA racemase
Formulation In PBS (0.02% sodium azide)
Gene AMACR
Gene Alias RACE
Gene Symbols AMACR
Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 33-48 of human AMACR.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23600
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.