Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AMACR Rabbit anti-Human, Polyclonal Antibody, Abnova™

Catalog No. 89121366 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-121-366 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-121-366 Supplier Abnova Corporation Supplier No. PAB19835
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of AMACR.

Alpha-methylacyl-CoA racemase is responsible for the conversion of pristanoyl-CoA and C27-bile acyl-CoAs to their (S)-stereoisomers, which are the only stereoisomers that can be degraded via peroxisomal beta-oxidation.[supplied by OMIM

Specifications

Antigen AMACR
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of AMACR.
Dilution ELISA (1:80000) Western Blot (1:200-1000) The optimal working dilution should be determined by the end user.
Formulation In serum (0.02% sodium azide)
Gene AMACR
Gene Accession No. Q9UHK6
Gene Alias RACE
Gene Symbols AMACR
Host Species Rabbit
Immunogen A synthetic peptide corresponding to 16 amino acids near internal region of human AMACR.
Purification Method Protein A purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23600
Target Species Human
Content And Storage Store at 4°C for three months. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.