Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Rabbit anti-AMACR, Polyclonal,

Catalog No. ABC925MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABC925MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABC925MI Supplier MilliporeSigma™ Supplier No. ABC925
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifically detects AMACR in Human samples, and it is validated for Immunohistochemistry, Western Blotting

Alpha-methylacyl-CoA racemase is encoded by the AMACR gene and catalyzes the convertion of various (2R)-methylacyl-CoA esters to the corresponding (2S)-methylacyl-CoA epimers. AMACR deficiency (AMACRD) is a rare autosomal recessive peroxisomal disorder characterized by elevated plasma concentrations of pristanic acid C27-bile-acid intermediates, resulting in variable neurodegenerative symptoms affecting the central and peripheral nervous systems, including seizures, visual failure, sensorimotor neuropathy, spasticity, migraine, and white matter hyperintensities on brain imaging.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AMACR
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Formulation Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. Q9UHK6
Gene Symbols AMACR
Host Species Rabbit
Immunogen GST-tagged recombinant protein corresponding to human AMACR near the N-terminal.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Apoptosis, Cancer
Primary or Secondary Primary
Gene ID (Entrez) NP_055139
Target Species Human
Content And Storage Stable for one year at 2°C to 8°C from date of receipt.
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.