Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AMBP Rabbit anti-Human, Polyclonal Antibody, Abnova™

Catalog No. 89113976 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-113-976 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-113-976 Supplier Abnova Corporation Supplier No. PAB0143
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against recombinant bikunin.

This gene encodes a complex glycoprotein secreted in plasma. The precursor is proteolytically processed into distinct functioning proteins: alpha-1-microglobulin, which belongs to the superfamily of lipocalin transport proteins and may play a role in the regulation of inflammatory processes, and bikunin, which is a urinary trypsin inhibitor belonging to the superfamily of Kunitz-type protease inhibitors and plays an important role in many physiological and pathological processes. This gene is located on chromosome 9 in a cluster of lipocalin genes. [provided by RefSeq

Specifications

Antigen AMBP
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against recombinant bikunin.
Dilution Western Blot (1:2000) ELISA (1:2000) The optimal working dilution should be determined by the end user.
Formulation Lyophilized from serum
Gene AMBP
Gene Alias EDC1/HCP/HI30/IATIL/ITI/ITIL/ITILC/UTI
Gene Symbols AMBP
Host Species Rabbit
Immunogen Recombinant protein corresponding to human bikunin.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 259
Target Species Human
Content And Storage Store at -20°C on dry atmosphere.
Aliquot to avoid repeated freezing and thawing.
Form Lyophilized
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.