Learn More
AMH, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of AMH.
Supplier: Abnova Corporation PAB19769
Description
Anti-Mullerian hormone is a member of the transforming growth factor-beta gene family which mediates male sexual differentiation. Anti-Mullerian hormone causes the regression of Mullerian ducts which would otherwise differentiate into the uterus and fallopian tubes. Some mutations in the anti-Mullerian hormone result in persistent Mullerian duct syndrome. [provided by RefSeq
Specifications
AMH | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of AMH. | |
In serum (0.02% sodium azide) | |
P03971 | |
AMH | |
A synthetic peptide corresponding to 16 amino acids near C-terminus of human AMH. | |
100 μg | |
Primary | |
Human | |
Liquid |
ELISA, Western Blot | |
Unconjugated | |
ELISA (1:10000-1:40000) Western Blot (1:200-1000) The optimal working dilution should be determined by the end user. | |
AMH | |
MIF/MIS | |
Rabbit | |
Protein A purification | |
RUO | |
268 | |
Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only