Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Amigo2, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89114867 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-114-867 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-114-867 Supplier Abnova Corporation Supplier No. PAB1072

Rabbit polyclonal antibody raised against synthetic peptide of Amigo2.

Sequence: TGDASADDRKAGKRVV

Specifications

Antigen Amigo2
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description adhesion molecule with Ig like domain 2
Formulation In PBS (0.02% sodium azide)
Gene Amigo2
Gene Alias AI415330, AW208913, Ali1
Gene Symbols Amigo2
Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 449-464 of mouse Amigo2.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 105827
Target Species Mouse
Content And Storage Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.