Learn More
Amyloid-beta Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against partial recombinant Amyloid-beta.
Supplier: Abnova Corporation PAB9713
Description
This gene encodes a cell surface receptor and transmembrane precursor protein that is cleaved by secretases to form a number of peptides. Some of these peptides are secreted and can bind to the acetyltransferase complex APBB1/TIP60 to promote transcriptional activation, while others form the protein basis of the amyloid plaques found in the brains of patients with Alzheimer disease. Mutations in this gene have been implicated in autosomal dominant Alzheimer disease and cerebroarterial amyloidosis (cerebral amyloid angiopathy). Multiple transcript variants encoding several different isoforms have been found for this gene. [provided by RefSeq
Specifications
Amyloid-beta | |
Polyclonal | |
amyloid beta (A4) precursor protein | |
APP | |
APP | |
Recombinant protein corresponding to amino acids 1-40 of human Amyloid-beta. | |
RUO | |
351 | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Dot Blot, ELISA, Immunohistochemistry, Radioimmune Assays (RIA) | |
Unconjugated | |
In PBS, pH 7.3 | |
AAA, ABETA, ABPP, AD1, APPI, CTFgamma, CVAP, PN2 | |
Rabbit | |
100 μg | |
Primary | |
Human | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only