Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

amyS, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89138436 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-138-436 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-138-436 Supplier Abnova Corporation Supplier No. PAB28061
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against native amyS (Bacillus amyloliquefaciens).

Specifications

Antigen amyS
Applications ELISA, Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against native amyS (Bacillus amyloliquefaciens).
Dilution ELISA (1:5000 - 1:20000) Immunoprecipitation (1:100) Western Blot (1:500-1:2000) The optimal working dilution should be determined by the end user.
Formulation In 20mM KH2PO4, 150mM NaCl, pH 7.2 (0.01% sodium azide)
Host Species Rabbit
Immunogen Native purified Bacillus amyloliquefaciens amyS.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7849308
Target Species Bacteria
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot after reconstitution to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.