Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ANAPC1 (phospho S377), Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89134507 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-134-507 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-134-507 Supplier Abnova Corporation Supplier No. PAB9936
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic phosphopeptide of ANAPC1.

ANAPC1 is 1 of at least 10 subunits of the anaphase-promoting complex (APC), which functions at the metaphase-to-anaphase transition of the cell cycle and is regulated by spindle checkpoint proteins. The APC is an E3 ubiquitin ligase that targets cell cycle regulatory proteins for degradation by the proteasome, thereby allowing progression through the cell cycle.[supplied by OMIM

Specifications

Antigen ANAPC1
Applications ELISA, Immunohistochemistry (PFA fixed), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic phosphopeptide of ANAPC1.
Dilution ELISA (1:2000-1:10000) Western Blot (1:200-1:1000) Immunohistochemistry (5.0 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In 20mM KH2PO4, 150mM NaCl, pH 7.2 (0.01% sodium azide)
Gene ANAPC1
Gene Accession No. Q9H1A4;NP_073153
Gene Alias APC1/MCPR/TSG24
Gene Symbols ANAPC1
Host Species Rabbit
Immunogen Synthetic phosphopeptide corresponding to residues surrounding S377 of human ANAPC1.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64682
Test Specificity This antibody is specific to phosphorylated human APC1 protein at the pS377 residue.
Target Species Bovine, Canine, Human, Mouse, Rat
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.