Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ANAPC11 Rabbit anti-Human, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89114563 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-114-563 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-114-563 Supplier Abnova Corporation Supplier No. PAB10020
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of ANAPC11.

Specifications

Antigen ANAPC11
Applications ELISA, Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of ANAPC11.
Dilution ELISA (1:2000-1:10000) Western Blot (1:500-1:1000) The optimal working dilution should be determined by the end user.
Formulation In antiserum (0.01% sodium azide)
Gene ANAPC11
Gene Accession No. Q9NYG5
Gene Alias APC11/Apc11p/HSPC214/MGC882
Gene Symbols ANAPC11
Host Species Rabbit
Immunogen A synthetic peptide (conjugated with KLH) corresponding to amino acids 76-84 of human ANAPC11.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51529
Test Specificity Cross reactivity may also occur with APC11 from other sources. Sufficient sequence differences exist to suggest that this antibody would not react with other RING box proteins such as ROC1 and ROC2.
Target Species Human, Mouse
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.