Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ANO1, Rabbit, Clone: SP31, Abnova™

Catalog No. 89110843 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-110-843 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-110-843 Supplier Abnova Corporation Supplier No. MAB2143

Rabbit monoclonal antibody raised against synthetic peptide of ANO1.

Specifications

Antigen ANO1
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone SP31
Conjugate Unconjugated
Description Rabbit monoclonal antibody raised against synthetic peptide of ANO1.
Dilution Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:100) The optimal working dilution should be determined by the end user.
Formulation Tissue culture supernatant in TBS buffer, pH 7.5 (1% BSA, 0.09% sodium azide).
Gene ANO1
Gene Alias FLJ10261/ORAOV2/TAOS2/TMEM16A
Gene Symbols ANO1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to human ANO1.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55107
Target Species Human
Content And Storage Store at 4°C. Do not freeze.
Form Liquid
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.