Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ANP32A Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™

Catalog No. 89116173 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-116-173 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-116-173 Supplier Abnova Corporation Supplier No. PAB12901
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of ANP32A.

Specifications

Antigen ANP32A
Applications Immunohistochemistry (PFA fixed), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of ANP32A.
Dilution Western Blot (0.5-2 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.02% sodium azide)
Gene ANP32A
Gene Accession No. P39687
Gene Alias C15orf1/I1PP2A/LANP/MAPM/MGC119787/MGC150373/PHAP1/PHAPI/PP32
Gene Symbols ANP32A
Host Species Rabbit
Immunogen A synthetic peptide corresponding to C-terminus of human ANP32A.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8125
Test Specificity A band at approximately 32 KDa can be detected. This polyclonal antibody has no cross-reaction to PHAP I2a and PHAP III.
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C for three months. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.