Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ANP32E, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89116172 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-116-172 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-116-172 Supplier Abnova Corporation Supplier No. PAB12900
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of ANP32E.

Specifications

Antigen ANP32E
Applications Immunohistochemistry (PFA fixed), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of ANP32E.
Dilution Western Blot (0.5-2 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.02% sodium azide)
Gene ANP32E
Gene Accession No. NP_112182
Gene Alias LANP-L/LANPL/MGC5350
Gene Symbols ANP32E
Host Species Rabbit
Immunogen A synthetic peptide corresponding to C-terminus of human ANP32E.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 81611
Test Specificity A band at approximately 35 KDa can be detected. PHAP III has no cross-reaction to PHAP I and PHAP I2a.
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C for three months. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.