Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ANXA11, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89121304 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-121-304 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-121-304 Supplier Abnova Corporation Supplier No. PAB19777
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of ANXA11.

This gene encodes a member of the annexin family, a group of calcium-dependent phospholipid-binding proteins. Annexins have unique N-terminal domains and conserved C-terminal domains, which contain the calcium-dependent phospholipid-binding sites. The encoded protein is a 56kDa antigen recognized by sera from patients with various autoimmune diseases. Transcript variants encoding the same isoform have been identified. [provided by RefSeq

Specifications

Antigen ANXA11
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of ANXA11.
Dilution ELISA (1:5000-1:20000) Western Blot (1:200-500) The optimal working dilution should be determined by the end user.
Formulation In serum (0.02% sodium azide)
Gene ANXA11
Gene Accession No. NP_001148
Gene Alias ANX11/CAP50
Gene Symbols ANXA11
Host Species Rabbit
Immunogen A synthetic peptide corresponding to 14 amino acids near N-terminus of human ANXA11.
Purification Method Protein A purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 311
Target Species Human
Content And Storage Store at 4°C for three months. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.