Learn More
ANXA6 Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of ANXA6.
Supplier: Abnova Corporation PAB19522
Description
Annexin VI belongs to a family of calcium-dependent membrane and phospholipid binding proteins. Although their functions are still not clearly defined, several members of the annexin family have been implicated in membrane-related events along exocytotic and endocytotic pathways. The annexin VI gene is approximately 60kb long and contains 26 exons. It encodes a protein of about 68kDa that consists of eight 68-amino acid repeats separated by linking sequences of variable lengths. It is highly similar to human annexins I and II sequences, each of which contain four such repeats. Exon 21 of annexin VI is alternatively spliced, giving rise to two isoforms that differ by a 6-amino acid insertion at the start of the seventh repeat. Annexin VI has been implicated in mediating the endosome aggregation and vesicle fusion in secreting epithelia during exocytosis. [provided by RefSeq
Specifications
ANXA6 | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of ANXA6. | |
Lyophilized from 0.9mg NaCl, 0.2mg Na2HPO4 (5mg BSA, 0.05mg sodium azide, 0.05mg Thimerosal) | |
ANX6/CBP68 | |
Rabbit | |
Immunoaffinity purification | |
RUO | |
309 | |
Human, Mouse, Rat | |
Lyophilized |
Western Blot | |
Unconjugated | |
Western Blot (1 ug/mL) The optimal working dilution should be determined by the end user. | |
ANXA6 | |
ANXA6 | |
A synthetic peptide corresponding to amino acids at C-terminus of human ANXA6. | |
100 μg | |
Primary | |
It identical to the related mouse and rat sequence. | |
Store at -20°C on dry atmosphere. After reconstitution with 200μL of deionized water and concentration will be 500μ/mL store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only