Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ANXA6 Rabbit anti-Human, Polyclonal Antibody, Abnova™

Catalog No. 89117311 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-117-311 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-117-311 Supplier Abnova Corporation Supplier No. PAB14863
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against full-length recombinant ANXA6.

Annexin VI belongs to a family of calcium-dependent membrane and phospholipid binding proteins. Although their functions are still not clearly defined, several members of the annexin family have been implicated in membrane-related events along exocytotic and endocytotic pathways. The annexin VI gene is approximately 60kb long and contains 26 exons. It encodes a protein of about 68kDa that consists of eight 68-amino acid repeats separated by linking sequences of variable lengths. It is highly similar to human annexins I and II sequences, each of which contain four such repeats. Exon 21 of annexin VI is alternatively spliced, giving rise to two isoforms that differ by a 6-amino acid insertion at the start of the seventh repeat. Annexin VI has been implicated in mediating the endosome aggregation and vesicle fusion in secreting epithelia during exocytosis. [provided by RefSeq

Specifications

Antigen ANXA6
Applications ELISA, Immunohistochemistry (PFA fixed), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against full length recombinant ANXA6.
Dilution Western Blot (1:1000) ELISA (1:1000-1:5000) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:1500) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4
Gene ANXA6
Gene Accession No. NM_001155.2
Gene Alias ANX6/CBP68
Gene Symbols ANXA6
Host Species Rabbit
Immunogen Recombinant 6xHis fusion protein corresponding to full length human ANXA6.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 309
Test Specificity Molecular mass ∼150000 da.
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.