Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AP1B1, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89117009 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-117-009 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-117-009 Supplier Abnova Corporation Supplier No. PAB14335
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit polyclonal antibody raised against synthetic peptide of AP1B1.

Adaptor protein complex 1 is found at the cytoplasmic face of coated vesicles located at the Golgi complex, where it mediates both the recruitment of clathrin to the membrane and the recognition of sorting signals within the cytosolic tails of transmembrane receptors. This complex is a heterotetramer composed of two large, one medium, and one small adaptin subunit. The protein encoded by this gene serves as one of the large subunits of this complex and is a member of the adaptin protein family. This gene is a candidate meningioma gene. Two transcript variants encoding different isoforms have been found for this gene, and variants utilizing alternative polyadenylation signals exist. [provided by RefSeq

Sequence: KKGEIFELKAELNSDKKEK

Specifications

Antigen AP1B1
Applications Immunohistochemistry (Frozen), Immunohistochemistry (PFA fixed), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of AP1B1.
Dilution Immunohistochemistry (1:1000-1:5000) Western Blot (1:100-1:10000) The optimal working dilution should be determined by the end user.
Formulation Lyophilized from PBS
Gene AP1B1
Gene Accession No. Q10567;O35643;P52303;Q2KJB2
Gene Alias ADTB1/AP105A/BAM22/CLAPB2
Gene Symbols AP1B1
Host Species Rabbit
Immunogen A synthetic peptide (conjugated with diphtheria toxoid) corresponding to amino acids 11-29 of human AP1B1.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 162
Test Specificity This antibody has been shown to be specific to AP-1 complex subunit beta-1.
Target Species Human, Rat
Content And Storage Store at 4°C on dry atmosphere. After reconstitution with deionized water store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.