Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AP3B2 Rabbit anti-Human, Polyclonal Antibody, Abnova™

Catalog No. 89125250 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-125-250 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-125-250 Supplier Abnova Corporation Supplier No. PAB23545
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against recombinant AP3B2.

Adaptor protein-3 (AP3) is a heterotetrameric vesicle-coat protein complex. Some AP3 subunits are ubiquitously expressed, whereas others are expressed exclusively in neurons. The neuron-specific AP3 complex, which includes AP3B2, is thought to serve neuron-specific functions such as neurotransmitter release (Grabner et al., 2006 [PubMed 16788073]).[supplied by OMIM

Sequence: APVFMSENEFKKEQGKLMGMNEITEKLMLPDTCRSDHIVVQKVTATANLGRVPCGTSDEYRFAGRTLTGGSLVLLTLDARPAGAAQLTVNSEKMVIGTMLVKDVIQ

Specifications

Antigen AP3B2
Applications Immunohistochemistry (PFA fixed)
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against recombinant AP3B2.
Dilution Immunohistochemistry (1:200-1:500) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide)
Gene AP3B2
Gene Accession No. Q13367
Gene Alias DKFZp686D17136/NAPTB
Gene Symbols AP3B2
Host Species Rabbit
Immunogen Recombinant protein corresponding to amino acids of human AP3B2.
Purification Method Antigen affinity purification
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8120
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.