Learn More
APH1A Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of APH1A.
Supplier: Abnova Corporation PAB8483
Description
APH1 is a multipass transmembrane protein that interacts with presenilin (see PSEN1; MIM 104311) and nicastrin (APH2; MIM 605254) as a functional component of the gamma-secretase complex. The gamma-secretase complex is required for the intramembrane proteolysis of a number of membrane proteins, including the amyloid-beta precursor protein (APP; MIM 104760) and Notch (MIM 190198).[supplied by OMIM
Specifications
APH1A | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of APH1A. | |
In PBS (0.1% proclin, 2.0% Block Ace) | |
6530402N02Rik/APH-1A/CGI-78 | |
Rabbit | |
25 μg | |
Primary | |
Human | |
Liquid |
Western Blot | |
Unconjugated | |
Western Blot (3-10 ug/mL) The optimal working dilution should be determined by the end user. | |
APH1A | |
APH1A | |
A synthetic peptide corresponding to human APH1A. | |
RUO | |
51107 | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only