Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aplnr, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89119619 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-119-619 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-119-619 Supplier Abnova Corporation Supplier No. PAB18032

Rabbit polyclonal antibody raised against synthetic peptide of Aplnr.

Specifications

Antigen Aplnr
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of Aplnr.
Dilution ELISA (0.1-1 ug/mL) Western Blot (1:1000-1:2000) The optimal working dilution should be determined by the end user.
Formulation Lyophilized from PBS.
Gene Aplnr
Gene Alias Agtrl1/Apj
Gene Symbols Aplnr
Host Species Rabbit
Immunogen A synthetic peptide corresponding to rat Aplnr.
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 83518
Test Specificity Rabbit polyclonal antibody to APLNR can detect cellular protein derived from cell lysates (3T3-L1, NIH-3T3) at various concentrations. Cross reactivity with human and mouse might be happened duo to their sequence similarity .
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C.
Aliquot after reconstitution to avoid repeated freezing and thawing.
Form Lyophilized
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.