Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
APLP2, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic phosphopeptide of human APLP2.
Supplier: Abnova Corporation PAB29234
Specifications
APLP2 | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic phosphopeptide of human APLP2. | |
In PBS, pH 7.4 (without mg2+ and Ca2+), (50% glycerol, 0.02% sodium azide) | |
Q06481 | |
APLP2 | |
Synthetic phosphopeptide (conjugated with KLH) corresponding to residues surrounding Y755 of human APLP2. | |
100 μL | |
Primary | |
This antibody detects endogenous levels of APLP2 only when phosphorylated at tyrosine 755 | |
Store at -20°C. Avoid repeated freezing and thawing. |
Immunohistochemistry (PFA fixed) | |
Unconjugated | |
Immunohistochemistry (1:50-1:100) The optimal working dilution should be determined by the end user. | |
APLP2 | |
APPH/APPL2/CDEBP | |
Rabbit | |
Affinity purification | |
RUO | |
334 | |
Human, Mouse, Rat | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction