Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Apoe, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89116873 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-116-873 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-116-873 Supplier Abnova Corporation Supplier No. PAB13892

Rabbit polyclonal antibody raised against native Apoe.

Specifications

Antigen Apoe
Applications Immunodiffusion, Radioimmune Assays (RIA), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against native Apoe.
Dilution Western Blot (1:500-1:1000) The optimal working dilution should be determined by the end user.
Formulation In buffer containing 0.05% sodium azide
Gene Apoe
Gene Alias AI255918
Gene Symbols Apoe
Host Species Rabbit
Immunogen Native purified Apoe from mouse plasma high and very low density lipoproteins.
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11816
Test Specificity Extremely low recognition of human Apo E. No data available regarding rat activity.
Target Species Human, Mouse
Content And Storage Store at -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.